missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, 95+%, for spectroscopy, mixture of isomers

Catalog No. AC168575000
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC168575000 500 mL Glass bottle
AC168570025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC168575000 Supplier Thermo Scientific Chemicals Supplier No. 168575000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 92112-69-1 | C6H14 | 86.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

Specifications

Chemical Name or Material Hexanes
Acidity 0.00015 meq/g max.
CAS 92112-69-1, 110-54-3
Absorbance (1 cm cell vs water), 0.005 max. at 240nm, 0.005 max. at 250nm, 0.01 max. at 230nm, 0.025 max. at 220nm, 0.1 max. at 210nm, 0.3 max. at 200nm
Melting Point -95°C
Density 0.6590g/mL
Boiling Point 69°C
Flash Point -22°C
Assay Percent Range (95% n-hexane approximately) (GC)
Packaging Glass bottle
Molecular Formula C6H14
Refractive Index 1.3748 to 1.381
Linear Formula CH3(CH2)4CH3
Quantity 500 mL
MDL Number MFCD00009520
Residue after Evaporation 0.0005% max.
Beilstein 01, 142
Specific Gravity 0.659
Merck Index 14, 4694
Synonym Hex
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,acetone,ether and chloroform
Molecular Weight (g/mol) 86.18
Viscosity 0.31 mPa.s (20°C)
Vapor Pressure 160mbar at 20°C
Formula Weight 86.18
Percent Purity 95+%
Grade Spectroscopy
Physical Form Liquid
Show More Show Less

RUO – Research Use Only