CAS RN 92028-57-4
IUPAC Name:
2-(2,5-dimethyl-1H-pyrrol-1-yl)benzoic acid
Synonyms:
2-(2,5-dimethylpyrrol-1-yl)benzoic acid
Molecular Weight (g/mol):
215.25
Molecular Formula:
C13H13NO2
InChi Key:
ZLYUUANOICYAAL-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C)N1C1=CC=CC=C1C(O)=O
1
–
3
of
3
results
2
2-(2,5-Dimethyl-1H-pyrrol-1-yl)benzoic acid, 97%, Thermo Scientific™
CAS: 92028-57-4 Molecular Formula: C13H13NO2 Molecular Weight (g/mol): 215.25 MDL Number: MFCD01125264 InChI Key: ZLYUUANOICYAAL-UHFFFAOYSA-N Synonym: 2-2,5-dimethyl-1h-pyrrol-1-yl benzoic acid,2-2,5-dimethylpyrrol-1-yl benzoic acid,2-2,5-dimethyl-1-pyrrolyl benzoic acid,2-2,5-dimethyl-pyrrol-1-yl-benzoic acid,2-2,5-dimethylpyrrolyl benzoic acid,cbmicro_048042,benzoic acid,2-2,5-dimethyl-1h-pyrrol-1-yl PubChem CID: 292956 IUPAC Name: 2-(2,5-dimethylpyrrol-1-yl)benzoic acid SMILES: CC1=CC=C(C)N1C1=CC=CC=C1C(O)=O