CAS RN 91872-10-5
CAS RN 91872-10-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Amino-3,5-dibromo-6-methylpyridine 98.0+%, TCI Americaâ„¢
CAS: 91872-10-5 Molecular Formula: C6H6Br2N2 Molecular Weight (g/mol): 265.936 MDL Number: MFCD00068229 InChI Key: BLUYMDMBXIMIGK-UHFFFAOYSA-N Synonym: 2-amino-3,5-dibromo-6-methylpyridine,3,5-dibromo-6-methyl-2-pyridinamine,2-amino-3,5-dibromo-6-picoline,6-amino-3,5-dibromo-2-picoline,3,5-dibromo-6-methyl-pyridin-2-ylamine,3,5-dibromo-6-methyl-2-pyridylamine,2-pyridinamine, 3,5-dibromo-6-methyl,wisteria sinensis extract,2-amino-3,5-dibrom-6-methylpyridine,pubchem7669 PubChem CID: 522994 IUPAC Name: 3,5-dibromo-6-methylpyridin-2-amine SMILES: CC1=NC(=C(C=C1Br)Br)N