CAS RN 916733-17-0
IUPAC Name:
1-(3-hydroxypyrrolidin-1-yl)ethan-1-one
Synonyms:
1-(3-hydroxypyrrolidin-1-yl)ethanone
Molecular Weight (g/mol):
129.16
Molecular Formula:
C6H11NO2
InChi Key:
ODHQVFPGHQBQSY-UHFFFAOYNA-N
SMILES:
CC(=O)N1CCC(O)C1
1
–
1
of
1
results
1
(R)-1-Acetyl-3-pyrrolidinol 98.0+%, TCI America™
CAS: 916733-17-0 Molecular Formula: C6H11NO2 Molecular Weight (g/mol): 129.159 InChI Key: ODHQVFPGHQBQSY-ZCFIWIBFSA-N Synonym: (R)-1-Acetyl-3-hydroxypyrrolidine PubChem CID: 10486876 IUPAC Name: 1-[(3R)-3-hydroxypyrrolidin-1-yl]ethanone SMILES: CC(=O)N1CCC(C1)O