CAS RN 91345-62-9
IUPAC Name:
1-[(4-bromophenyl)methyl]piperazine
Synonyms:
1-[(4-bromophenyl)methyl]piperazine
Molecular Weight (g/mol):
255.16
Molecular Formula:
C11H15BrN2
InChi Key:
MAHWBNAOEVAPJF-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(CN2CCNCC2)C=C1
1
–
1
of
1
results
1
Promotions Available
1-(4-Bromobenzyl)piperazine, 97%, Thermo Scientific™
1-(4-Bromobenzyl)piperazine, 97%, Thermo Scientific™
CAS: 91345-62-9 Molecular Formula: C11H15BrN2 Molecular Weight (g/mol): 255.159 MDL Number: MFCD02177416 InChI Key: MAHWBNAOEVAPJF-UHFFFAOYSA-N Synonym: 1-4-bromobenzyl piperazine,1-4-bromophenyl methyl piperazine,1-4-bromo-benzyl-piperazine,piperazine, 1-4-bromophenyl methyl,cbmicro_035222,acmc-20apb8,cambridge id 5862395,1-4-bromobenzyl-piperazine,4-bromophenyl methyl piperazine PubChem CID: 876494 IUPAC Name: 1-[(4-bromophenyl)methyl]piperazine SMILES: C1CN(CCN1)CC2=CC=C(C=C2)Br