CAS RN 91328-77-7
IUPAC Name:
4-ethylcyclohexane-1-carboxylic acid
Synonyms:
4-ethylcyclohexane-1-carboxylic acid
Molecular Weight (g/mol):
156.23
Molecular Formula:
C9H16O2
InChi Key:
UNROFSAOTBVBBT-UHFFFAOYSA-N
SMILES:
CCC1CCC(CC1)C(O)=O
1
–
1
of
1
results
1
4-Ethylcyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™
CAS: 91328-77-7 Molecular Formula: C9H16O2 Molecular Weight (g/mol): 156.23 MDL Number: MFCD01076323,MFCD01568857 InChI Key: UNROFSAOTBVBBT-UHFFFAOYSA-N Synonym: trans-4-ethylcyclohexanecarboxylic acid,4-ethylcyclohexanecarboxylic acid,cyclohexanecarboxylic acid, 4-ethyl,4-trans-ethyl cyclohexane carboxylic acid,cyclohexanecarboxylic acid, 4-ethyl-, trans,1s,4r-4-ethylcyclohexane-1-carboxylic acid,pubchem2491,acmc-209o2z,ksc352s1t,ksc352s2b PubChem CID: 1490288 IUPAC Name: 4-ethylcyclohexane-1-carboxylic acid SMILES: CCC1CCC(CC1)C(O)=O