accessibility menu, dialog, popup

UserName

BOCH3FFFK

CAS RN 910251-11-5

IUPAC Name: potassium trifluoro(methoxymethyl)boranuide
Synonyms: potassium trifluoro(methoxymethyl)boranuide
Molecular Weight (g/mol): 151.96
Molecular Formula: C2H5BF3KO
InChi Key: WNVXSBCLLHGATG-UHFFFAOYSA-N
SMILES: [K+].COC[B-](F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results