CAS RN 910037-04-6
IUPAC Name:
dimethyl(3-{4-[(methylamino)methyl]phenoxy}propyl)amine
Synonyms:
dimethyl(3-{4-[(methylamino)methyl]phenoxy}propyl)amine
Molecular Weight (g/mol):
222.33
Molecular Formula:
C13H22N2O
InChi Key:
LFLFEMOINFHORX-UHFFFAOYSA-N
SMILES:
CNCC1=CC=C(OCCCN(C)C)C=C1
1
–
1
of
1
results
1
4-[3-(Dimethylamino)propoxy]-N-methylbenzylamine, 90%, Thermo Scientific™
CAS: 910037-04-6 Molecular Formula: C13H22N2O Molecular Weight (g/mol): 222.332 MDL Number: MFCD09064994 InChI Key: LFLFEMOINFHORX-UHFFFAOYSA-N Synonym: 4-3-dimethylamino propoxy-n-methylbenzylamine,dimethyl 3-4-methylamino methyl phenoxy propyl amine,n,n-dimethyl-3-4-methylamino methyl phenoxy propan-1-amine,4-3-dimethylamino propoxy phenyl methyl methyl amine,benzenemethanamine, 4-3-dimethylamino propoxy-n-methyl PubChem CID: 24229610 IUPAC Name: N,N-dimethyl-3-[4-(methylaminomethyl)phenoxy]propan-1-amine SMILES: CNCC1=CC=C(C=C1)OCCCN(C)C