accessibility menu, dialog, popup

UserName

CAS RN 90897-92-0

IUPAC Name: bromo(3,4-difluorophenyl)magnesium
Synonyms: bromo(3,4-difluorophenyl)magnesium
Molecular Weight (g/mol): 217.3
Molecular Formula: C6H3BrF2Mg
InChi Key: WZHAVYGMKGPNFN-UHFFFAOYSA-M
SMILES: FC1=CC=C([Mg]Br)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results