CAS RN 906352-99-6
IUPAC Name:
ethyl 3-[(6-methylpyrazin-2-yl)oxy]benzoate
Synonyms:
ethyl 3-[(6-methylpyrazin-2-yl)oxy]benzoate
Molecular Weight (g/mol):
258.28
Molecular Formula:
C14H14N2O3
InChi Key:
ALDBUDYDJSLBJO-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC(OC2=NC(C)=CN=C2)=CC=C1
1
–
2
of
2
results
1
Ethyl 3-[(6-methylpyrazin-2-yl)oxy]benzoate, 97%, Thermo Scientific™
CAS: 906352-99-6 Molecular Formula: C14H14N2O3 Molecular Weight (g/mol): 258.28 MDL Number: MFCD09702407 InChI Key: ALDBUDYDJSLBJO-UHFFFAOYSA-N Synonym: ethyl 3-6-methylpyrazin-2-yl oxy benzoate,ethyl 3-6-methylpyrazin-2-yloxy benzoate,benzoic acid,3-6-methyl-2-pyrazinyl oxy-,ethyl ester PubChem CID: 24229683 IUPAC Name: ethyl 3-(6-methylpyrazin-2-yl)oxybenzoate SMILES: CCOC(=O)C1=CC(OC2=NC(C)=CN=C2)=CC=C1