accessibility menu, dialog, popup

UserName

CH3NNFFFBr

CAS RN 906352-89-4

IUPAC Name: 5-(2-bromophenyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole
Synonyms: 5-(2-bromophenyl)-1-methyl-3-(trifluoromethyl)pyrazole
Molecular Weight (g/mol): 305.1
Molecular Formula: C11H8BrF3N2
InChi Key: UKDUPCWWWMHDRH-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1C1=CC=CC=C1Br)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)