accessibility menu, dialog, popup

UserName

CAS RN 902518-11-0

NBr(?)

CAS RN 902518-11-0

IUPAC Name: 9-(2-bromophenyl)-9H-carbazole
Synonyms: 9-(2-bromophenyl)carbazole
Molecular Weight (g/mol): 322.21
Molecular Formula: C18H12BrN
InChi Key: KEWDVYIULXXMPP-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1N1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results