accessibility menu, dialog, popup

UserName

CAS RN 9007-43-6

IUPAC Name: (1Z,6Z,17Z)-15,19-bis(2-carboxyethyl)-4,9-bis[1-(cysteinylsulfanyl)ethyl]-5,10,14,20-tetramethyl-21,23,24,25-tetraaza-22-ferrahexacyclo[9.9.3.1³,⁶.1¹³,¹⁶.0⁸,²³.0¹⁸,²¹]pentacosa-1,3(25),4,6,8,10,14,16(24),17,19-decaene-22,22-bis(ylium)
Synonyms: (1Z,6Z,17Z)-15,19-bis(2-carboxyethyl)-4,9-bis[1-(cysteinylsulfanyl)ethyl]-5,10,14,20-tetramethyl-21,23,24,25-tetraaza-22-ferrahexacyclo[9.9.3.1³,⁶.1¹³,¹⁶.0⁸,²³.0¹⁸,²¹]pentacosa-1,3(25),4,6,8,10,14,16(24),17,19-decaene-22,22-bis(ylium)
Molecular Weight (g/mol): 892.94
Molecular Formula: C40H48FeN6O6S4
InChi Key: UMUOLVQYJUIZKD-RHANTFMNSA-L
SMILES: CC(SC(=O)[C@@H](N)CS)C1=C(C)\C2=C\C3=C(C(C)SC(=O)[C@@H](N)CS)C(C)=C4CC5N=C(\C=C6/N([Fe++]N34)\C(=C/C1=N2)C(C)=C6CCC(O)=O)C(CCC(O)=O)=C5C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results