accessibility menu, dialog, popup

UserName

BOH3CFHOOH

CAS RN 900174-60-9

IUPAC Name: (5-ethoxy-2-fluorophenyl)boronic acid
Synonyms: 5-ethoxy-2-fluorophenylboronic acid
Molecular Weight (g/mol): 183.97
Molecular Formula: C8H10BFO3
InChi Key: ISJGNUFTQTZLEZ-UHFFFAOYSA-N
SMILES: CCOC1=CC(B(O)O)=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results