CAS RN 89530-34-7
IUPAC Name:
1-phenyl-3-(trimethylsilyl)prop-2-yn-1-ol
Synonyms:
1-phenyl-3-(trimethylsilyl)prop-2-yn-1-ol
Molecular Weight (g/mol):
204.34
Molecular Formula:
C12H16OSi
InChi Key:
RVTDALNDYLDVMN-UHFFFAOYNA-N
SMILES:
C[Si](C)(C)C#CC(O)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
1-Phenyl-3-trimethylsilyl-2-propyn-1-ol, 97%
1-Phenyl-3-trimethylsilyl-2-propyn-1-ol, 97%
CAS: 89530-34-7 Molecular Formula: C12H16OSi Molecular Weight (g/mol): 204.344 MDL Number: MFCD05864336 InChI Key: RVTDALNDYLDVMN-UHFFFAOYSA-N Synonym: 1-phenyl-3-trimethylsilyl-2-propyn-1-ol,1-phenyl-3-trimethylsilyl prop-2-yn-1-ol,benzenemethanol, a-2-trimethylsilyl ethynyl,acmc-20ln8h,alpha-trimethylsilyl ethynylbenzyl alcohol,4,4-dimethyl-1-phenyl-4-silapent-2-yn-1-ol,alpha-trimethylsilyl ethynyl-benzenemethanol,alpha-trimethylsilyl-ethynyl-benzenemethanol,alpha-trimethylsilyl ethynyl-benzene-methanol,1-phenyl-3-1,1,1-trimethylsilyl-2-propyn-1-ol PubChem CID: 2760416 IUPAC Name: 1-phenyl-3-trimethylsilylprop-2-yn-1-ol SMILES: C[Si](C)(C)C#CC(C1=CC=CC=C1)O