accessibility menu, dialog, popup

UserName

CAS RN 894-71-3

IUPAC Name: hydrogen methyl(3-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-ylidene}propyl)amine chloride
Synonyms: hydrogen nortriptyline chloride
Molecular Weight (g/mol): 299.84
Molecular Formula: C19H22ClN
InChi Key: SHAYBENGXDALFF-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CNCCC=C1C2=CC=CC=C2CCC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results