accessibility menu, dialog, popup

UserName

CAS RN 89-39-4

H3COOCH3NOO

CAS RN 89-39-4

IUPAC Name: 1,4-dimethoxy-2-nitrobenzene
Synonyms: 1,4-dimethoxy-2-nitrobenzene
Molecular Weight (g/mol): 183.16
Molecular Formula: C8H9NO4
InChi Key: UPTOWXNJLZJTGD-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results