accessibility menu, dialog, popup

UserName

H3COOF

CAS RN 88754-96-5

IUPAC Name: 6-fluoro-2-methyl-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: 6-fluoro-2-methyl-2,3-dihydro-1-benzopyran-4-one
Molecular Weight (g/mol): 180.18
Molecular Formula: C10H9FO2
InChi Key: RPAIBTVEPAACRP-UHFFFAOYNA-N
SMILES: CC1CC(=O)C2=C(O1)C=CC(F)=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results