accessibility menu, dialog, popup

UserName

H3COONH

CAS RN 882-36-0

IUPAC Name: N-benzyl-3-oxobutanamide
Synonyms: N-benzyl-3-oxobutanamide
Molecular Weight (g/mol): 191.23
Molecular Formula: C11H13NO2
InChi Key: KOHNUEXAOQRRPI-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)NCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results