accessibility menu, dialog, popup

UserName

CAS RN 88043-21-4

CH3SOOONOOH3CH3CCH3OOCH3

CAS RN 88043-21-4

IUPAC Name: 1-tert-butyl 2-methyl 4-[(4-methylbenzenesulfonyl)oxy]pyrrolidine-1,2-dicarboxylate
Synonyms: 1-tert-butyl 2-methyl 4-[(4-methylbenzenesulfonyl)oxy]pyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 399.46
Molecular Formula: C18H25NO7S
InChi Key: UKVKNGDXVREPDE-UHFFFAOYNA-N
SMILES: COC(=O)C1CC(CN1C(=O)OC(C)(C)C)OS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results