accessibility menu, dialog, popup

UserName

NH2ClClSOHOO

CAS RN 88-50-6

IUPAC Name: 4-amino-2,5-dichlorobenzene-1-sulfonic acid
Synonyms: 4-amino-2,5-dichlorobenzenesulfonic acid
Molecular Weight (g/mol): 242.07
Molecular Formula: C6H5Cl2NO3S
InChi Key: SJCTXIKOXTUQHC-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=C(C=C1Cl)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results