accessibility menu, dialog, popup

UserName

NNOOHFFF

CAS RN 876728-42-6

IUPAC Name: 4-[2-(trifluoromethyl)-1H-1,3-benzodiazol-1-yl]butanoic acid
Synonyms: 4-[2-(trifluoromethyl)-1,3-benzodiazol-1-yl]butanoic acid
Molecular Weight (g/mol): 272.23
Molecular Formula: C12H11F3N2O2
InChi Key: HQUSGKZLJZMIHU-UHFFFAOYSA-N
SMILES: OC(=O)CCCN1C2=CC=CC=C2N=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)