CAS RN 876-53-9
IUPAC Name:
1,3-dibromoadamantane
Synonyms:
1,3-dibromoadamantane
Molecular Weight (g/mol):
294.03
Molecular Formula:
C10H14Br2
InChi Key:
HLWZKLMEOVIWRK-UHFFFAOYSA-N
SMILES:
BrC12CC3CC(C1)CC(Br)(C3)C2
1
–
1
of
1
results
1
1,3-Dibromoadamantane 97.0+%, TCI America™
CAS: 876-53-9 Molecular Formula: C10H14Br2 Molecular Weight (g/mol): 294.03 MDL Number: MFCD00077218 InChI Key: HLWZKLMEOVIWRK-UHFFFAOYSA-N PubChem CID: 265790 IUPAC Name: 1,3-dibromoadamantane SMILES: C1C2CC3(CC1CC(C2)(C3)Br)Br