accessibility menu, dialog, popup

UserName

CAS RN 875781-17-2

IUPAC Name: 5-bromo-1H-pyrazolo[3,4-b]pyridine
Synonyms: 5-bromo-1H-pyrazolo[3,4-b]pyridine
Molecular Weight (g/mol): 198.02
Molecular Formula: C6H4BrN3
InChi Key: BASYLPMLKGQZOG-UHFFFAOYSA-N
SMILES: BrC1=CN=C2NN=CC2=C1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)