CAS RN 873566-75-7
IUPAC Name:
(3-amino-4-fluorophenyl)boronic acid
Synonyms:
3-amino-4-fluorophenylboronic acid
Molecular Weight (g/mol):
154.94
Molecular Formula:
C6H7BFNO2
InChi Key:
SYBMNJPUZMUPGQ-UHFFFAOYSA-N
SMILES:
NC1=C(F)C=CC(=C1)B(O)O
1
–
1
of
1
results
1
Promotions Available
3-Amino-4-fluorophenylboronic acid, 97%
3-Amino-4-fluorophenylboronic acid, 97%
CAS: 873566-75-7 Molecular Formula: C6H7BFNO2 Molecular Weight (g/mol): 154.94 MDL Number: MFCD03092925 InChI Key: SYBMNJPUZMUPGQ-UHFFFAOYSA-N Synonym: 3-amino-4-fluorophenyl boronic acid,3-amino-4-fluorobenzeneboronic acid,5-borono-2-fluoroaniline,3-amino-4-fluoro-phenyl boronic acid,boronic acid, 3-amino-4-fluorophenyl,boronic acid,b-3-amino-4-fluorophenyl,hydrochloride,pubchem1743,acmc-209qjr,3-amino-4-fluoro phenylboronic acid PubChem CID: 21954814 IUPAC Name: (3-amino-4-fluorophenyl)boronic acid SMILES: NC1=C(F)C=CC(=C1)B(O)O