CAS RN 870-93-9
IUPAC Name:
2-amino-4-[(3-amino-3-carboxypropyl)disulfanyl]butanoic acid
Synonyms:
homocystine L-homocystine 4,4'-dithiobis DL-homocystine homocystine, L-
Molecular Weight (g/mol):
268.35
Molecular Formula:
C8H16N2O4S2
InChi Key:
ZTVZLYBCZNMWCF-UHFFFAOYNA-N
SMILES:
NC(CCSSCCC(N)C(O)=O)C(O)=O
1
–
1
of
1
results
1
DL-Homocystine 98.0+%, TCI America™
CAS: 870-93-9 Molecular Formula: C8H16N2O4S2 Molecular Weight (g/mol): 268.346 MDL Number: MFCD00069587 InChI Key: ZTVZLYBCZNMWCF-UHFFFAOYSA-N Synonym: dl-homocystine,homocystine,4,4'-disulfanediylbis 2-aminobutanoic acid,l-homocystine,oxidized dl-homocysteine,4,4'-dithiobis 2-aminobutyric acid,d-homocystine,2-amino-4-3-amino-3-carboxypropyl disulfanyl butanoic acid,butanoic acid, 4,4'-dithiobis 2-amino,butyric acid,4'-dithiobis 2-amino PubChem CID: 10010 ChEBI: CHEBI:17485 IUPAC Name: 2-amino-4-[(3-amino-3-carboxypropyl)disulfanyl]butanoic acid SMILES: C(CSSCCC(C(=O)O)N)C(C(=O)O)N