accessibility menu, dialog, popup

UserName

OOOOOOBr

CAS RN 86610-66-4

IUPAC Name: 4-{bis[4-(benzoyloxy)phenyl](bromo)methyl}phenyl benzoate
Synonyms: 4-{bis[4-(benzoyloxy)phenyl](bromo)methyl}phenyl benzoate
Molecular Weight (g/mol): 683.55
Molecular Formula: C40H27BrO6
InChi Key: PKXTVZCJOBWKIG-UHFFFAOYSA-N
SMILES: BrC(C1=CC=C(OC(=O)C2=CC=CC=C2)C=C1)(C1=CC=C(OC(=O)C2=CC=CC=C2)C=C1)C1=CC=C(OC(=O)C2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results