accessibility menu, dialog, popup

UserName

NNNBr

CAS RN 865156-68-9

IUPAC Name: 6-bromoimidazo[1,2-a]pyrimidine
Synonyms: 6-bromoimidazo[1,2-a]pyrimidine
Molecular Weight (g/mol): 198.02
Molecular Formula: C6H4BrN3
InChi Key: BQMWMOQCMFLRQQ-UHFFFAOYSA-N
SMILES: BrC1=CN2C=CN=C2N=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results