accessibility menu, dialog, popup

UserName

CAS RN 86-79-3

IUPAC Name: 9H-carbazol-2-ol
Synonyms: 2-hydroxycarbazole
Molecular Weight (g/mol): 183.21
Molecular Formula: C12H9NO
InChi Key: GWPGDZPXOZATKL-UHFFFAOYSA-N
SMILES: OC1=CC2=C(C=C1)C1=C(N2)C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results