CAS RN 86-65-7
CAS RN 86-65-7
IUPAC Name:
7-aminonaphthalene-1,3-disulfonic acid
Synonyms:
7-aminonaphthalene-1,3-disulfonic acid
Molecular Weight (g/mol):
303.3
Molecular Formula:
C10H9NO6S2
InChi Key:
CMOLPZZVECHXKN-UHFFFAOYSA-N
SMILES:
NC1=CC=C2C=C(C=C(C2=C1)S(O)(=O)=O)S(O)(=O)=O
1
–
3
of
3
results
3
7-Amino-1,3-naphthalenedisulfonic acid, Tech.
CAS: 86-65-7 Molecular Formula: C10H9NO6S2 Molecular Weight (g/mol): 303.29 MDL Number: MFCD00003991 InChI Key: CMOLPZZVECHXKN-UHFFFAOYSA-N Synonym: amido-g-acid,7-amino-1,3-naphthalenedisulfonic acid,amino-g-acid,1,3-naphthalenedisulfonic acid, 7-amino,amino-g acid,2-amino-6,8-disulfonaphthalene,unii-7c482fx29k,2-aminonaphthalene-6,8-disulfonic acid,2-naphthylamine-6,8-disulfonic acid,beta-naphthylamine-6,8-disulfonic acid PubChem CID: 6851 IUPAC Name: 7-aminonaphthalene-1,3-disulfonic acid SMILES: C1=CC(=CC2=C(C=C(C=C21)S(=O)(=O)O)S(=O)(=O)O)N