accessibility menu, dialog, popup

UserName

CAS RN 857283-91-1

IUPAC Name: 3-[(morpholin-4-yl)methyl]benzonitrile
Synonyms: 3-(morpholin-4-ylmethyl)benzonitrile
Molecular Weight (g/mol): 202.26
Molecular Formula: C12H14N2O
InChi Key: GSVNKQLSALKJHW-UHFFFAOYSA-N
SMILES: N#CC1=CC(CN2CCOCC2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results