accessibility menu, dialog, popup

UserName

FFOO

CAS RN 85684-61-3

IUPAC Name: 3-(difluoromethoxy)benzaldehyde
Synonyms: 3-(difluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: HFIUSWPRDIPIPN-UHFFFAOYSA-N
SMILES: FC(F)OC1=CC=CC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results