accessibility menu, dialog, popup

UserName

ClNNCl

CAS RN 85331-33-5

IUPAC Name: 3,5-dichloropyridine-2-carbonitrile
Synonyms: 3,5-dichloropyridine-2-carbonitrile
Molecular Weight (g/mol): 173
Molecular Formula: C6H2Cl2N2
InChi Key: ATUOLSDAAPMVJJ-UHFFFAOYSA-N
SMILES: ClC1=CN=C(C#N)C(Cl)=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)