accessibility menu, dialog, popup

UserName

NClFFF

CAS RN 85148-26-1

IUPAC Name: 3-chloro-5-(trifluoromethyl)pyridine
Synonyms: 3-chloro-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 181.54
Molecular Formula: C6H3ClF3N
InChi Key: OMRCXTBFBBWTDL-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=CN=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results