accessibility menu, dialog, popup

UserName

CH3NNH3CClFeClClCl

CAS RN 850331-04-3

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium; tetrachloroironuide
Synonyms: 1-ethyl-3-methylimidazolium; tetrachloroferrate(1-)
Molecular Weight (g/mol): 308.81
Molecular Formula: C6H11Cl4FeN2
InChi Key: VGSZFQMHQHFXCD-UHFFFAOYSA-J
SMILES: Cl[Fe-](Cl)(Cl)Cl.CCN1C=C[N+](C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results