accessibility menu, dialog, popup

UserName

CAS RN 85-15-4

H3CON

CAS RN 85-15-4

IUPAC Name: 2-methylnaphtho[1,2-d][1,3]oxazole
Synonyms: 2-methylnaphtho[1,2-d][1,3]oxazole
Molecular Weight (g/mol): 183.21
Molecular Formula: C12H9NO
InChi Key: JCTNVNANPZAULC-UHFFFAOYSA-N
SMILES: CC1=NC2=C(O1)C=CC1=CC=CC=C21

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results