accessibility menu, dialog, popup

UserName

CAS RN 84371-65-3

IUPAC Name: (1S,3aS,3bS,10R,11aS)-10-[4-(dimethylamino)phenyl]-1-hydroxy-11a-methyl-1-(prop-1-yn-1-yl)-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: mifepristone mifeprex mifegyne mifepristonum mifégyne mifepristone (substance) mifepristona corlux mifepristone danco brand exelgyn brand of mifepristone
Molecular Weight (g/mol): 429.6
Molecular Formula: C29H35NO2
InChi Key: VKHAHZOOUSRJNA-GCNJZUOMSA-N
SMILES: CC#C[C@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CCC4=C3[C@H](C[C@]12C)C1=CC=C(C=C1)N(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)