accessibility menu, dialog, popup

UserName

CH3NONNNHFOOHCH3NONNNHFOOHH2O

CAS RN 84294-96-2

IUPAC Name: bis(1-ethyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid) hydrate
Synonyms: bis(enoxacin) hydrate
Molecular Weight (g/mol): 658.66
Molecular Formula: C30H36F2N8O7
InChi Key: KIJBMNCGVILNLN-UHFFFAOYSA-N
SMILES: O.CCN1C=C(C(O)=O)C(=O)C2=CC(F)=C(N=C12)N1CCNCC1.CCN1C=C(C(O)=O)C(=O)C2=CC(F)=C(N=C12)N1CCNCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results