accessibility menu, dialog, popup

UserName

CAS RN 84228-93-3

IUPAC Name: 3-(pyridin-4-yl)prop-2-enoic acid
Synonyms: 3-(pyridin-4-yl)prop-2-enoic acid
Molecular Weight (g/mol): 149.15
Molecular Formula: C8H7NO2
InChi Key: SSAYTINUCCRGDR-UHFFFAOYSA-N
SMILES: OC(=O)C=CC1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results