accessibility menu, dialog, popup

UserName

CAS RN 841251-37-4

H3CNH3CFFFSOONSOOFFF

CAS RN 841251-37-4

IUPAC Name: 1-ethyl-3-methylpyridin-1-ium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: 1-ethyl-3-methylpyridin-1-ium; bistriflylimide anion
Molecular Weight (g/mol): 402.33
Molecular Formula: C10H12F6N2O4S2
InChi Key: UZIAGXMMNLATPP-UHFFFAOYSA-N
SMILES: CC[N+]1=CC=CC(C)=C1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results