accessibility menu, dialog, popup

UserName

CAS RN 83803-80-9

IUPAC Name: 1-butyl-4-(diethoxymethyl)benzene
Synonyms: 1-butyl-4-(diethoxymethyl)benzene
Molecular Weight (g/mol): 236.36
Molecular Formula: C15H24O2
InChi Key: VZIJDIWCWDYVFM-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=C1)C(OCC)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results