accessibility menu, dialog, popup

UserName

CAS RN 83204-68-6

IUPAC Name: 2,9-dibromo-6,13-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1,3,8,10,15-pentaene-5,7,12,14-tetrone
Synonyms: 2,9-dibromo-6,13-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1,3,8,10,15-pentaene-5,7,12,14-tetrone
Molecular Weight (g/mol): 425.97
Molecular Formula: C14H2Br2O6
InChi Key: FEDCHNPLDKDTNS-UHFFFAOYSA-N
SMILES: BrC1=C2C(=O)OC(=O)C3=CC(Br)=C4C(=O)OC(=O)C(=C1)C4=C23
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results