man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 830-50-2

H3COHFFFFF

CAS RN 830-50-2

IUPAC Name: 1-(2,3,4,5,6-pentafluorophenyl)ethan-1-ol
Synonyms: 1-(2,3,4,5,6-pentafluorophenyl)ethanol
Molecular Weight (g/mol): 212.12
Molecular Formula: C8H5F5O
InChi Key: WYUNHWKTLDBPLE-UHFFFAOYNA-N
SMILES: CC(O)C1=C(F)C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results