CAS RN 83-49-8
IUPAC Name:
(4R)-4-[(1R,3aS,3bS,5S,5aR,7R,9aR,9bS,11aR)-5,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Synonyms:
hyodeoxycholic acid|hyodesoxycholic acid|7-deoxyhyocholic acid|6α-hydroxylithocholate
Molecular Weight (g/mol):
392.58
Molecular Formula:
C24H40O4
InChi Key:
DGABKXLVXPYZII-SIBKNCMHSA-N
SMILES:
[H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])C[C@H](O)[C@]2([H])C[C@H](O)CC[C@]12C
1
–
1
of
1
results
1
Promotions Available
Hyodeoxycholic acid, 96%, Thermo Scientific Chemicals
Hyodeoxycholic acid, 96%, Thermo Scientific Chemicals
CAS: 83-49-8 Molecular Formula: C24H40O4 Molecular Weight (g/mol): 392.58 MDL Number: MFCD00003681 InChI Key: DGABKXLVXPYZII-SIBKNCMHSA-N Synonym: hyodeoxycholic acid PubChem CID: 124303820 IUPAC Name: (4S)-4-[(3S,5S,6R,8R,9R,10S,13S,14R,17S)-3,6-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid SMILES: [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])C[C@H](O)[C@]2([H])C[C@H](O)CC[C@]12C