accessibility menu, dialog, popup

UserName

CH3NSOOONa

CAS RN 82611-85-6

IUPAC Name: sodium 3-[ethyl(phenyl)amino]propane-1-sulfonate
Synonyms: sodium 3-[ethyl(phenyl)amino]propane-1-sulfonate
Molecular Weight (g/mol): 265.3
Molecular Formula: C11H16NNaO3S
InChi Key: FFJBIXKLISICDT-UHFFFAOYSA-M
SMILES: [Na+].CCN(CCCS([O-])(=O)=O)C1=CC=CC=C1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results