accessibility menu, dialog, popup

UserName

S

CAS RN 825-55-8

IUPAC Name: 2-phenylthiophene
Synonyms: 2-phenylthiophene
Molecular Weight (g/mol): 160.23
Molecular Formula: C10H8S
InChi Key: PJRGDKFLFAYRBV-UHFFFAOYSA-N
SMILES: S1C=CC=C1C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)