accessibility menu, dialog, popup

UserName

ClClSOOCl

CAS RN 82417-45-6

IUPAC Name: 2,3-dichlorobenzene-1-sulfonyl chloride
Synonyms: 2,3-dichlorobenzenesulfonyl chloride
Molecular Weight (g/mol): 245.5
Molecular Formula: C6H3Cl3O2S
InChi Key: PQODWTNHDKDHIW-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(=C1Cl)S(Cl)(=O)=O
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)