accessibility menu, dialog, popup

UserName

ClCH3PH3CCH3N

CAS RN 82358-61-0

IUPAC Name: tributyl(cyanomethyl)phosphanium chloride
Synonyms: tributyl(cyanomethyl)phosphanium chloride
Molecular Weight (g/mol): 277.82
Molecular Formula: C14H29ClNP
InChi Key: JCHWNYYARSRCKZ-UHFFFAOYSA-M
SMILES: [Cl-].CCCC[P+](CCCC)(CCCC)CC#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results