accessibility menu, dialog, popup

UserName

H3COCH3O

CAS RN 822-20-8

IUPAC Name: heptadecyl acetate
Synonyms: heptadecyl acetate
Molecular Weight (g/mol): 298.51
Molecular Formula: C19H38O2
InChi Key: BGBUNTANUVVMIQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCOC(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results