accessibility menu, dialog, popup

UserName

CAS RN 81971-39-3

CH3NOBr

CAS RN 81971-39-3

IUPAC Name: 5-bromo-1-methyl-1,2-dihydropyridin-2-one
Synonyms: 5-bromo-1-methylpyridin-2-one
Molecular Weight (g/mol): 188.02
Molecular Formula: C6H6BrNO
InChi Key: HHVFVHIUOMMWRN-UHFFFAOYSA-N
SMILES: CN1C=C(Br)C=CC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results